EMAIL THIS PAGE TO A FRIEND
Epilepsy currents
Hippocampal cell loss in posttraumatic human epilepsy.
PMID 18049724
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse