EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Asymmetric synthesis of (+)-cis-nemorensic acid from a chiral Diels-Alder adduct of 2,5-dimethylfuran.
PMID 18049754
Abstract
(+)-cis-Nemorensic acid was synthesized from a chiral Diels-Alder adduct prepared by a catalytic enantioselective Diels-Alder reaction with 2,5-dimethylfuran and 2,2,2-trifluoroethyl acrylate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
177717
2,5-Dimethylfuran, 99%
C6H8O