EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[A mammalian histone deacetylase related to the yeast transcriptional regulator Rpd3p].
PMID 18051428
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2580 Trapoxin A, ≥98% (HPLC), from Helicoma ambiens
C34H42N4O6