EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on triterpenoid constituents in leaf of Ilex pernyi].
PMID 18051898
Abstract
To investigate the chemical constituents of Ilex pernyi. The chemical constituents were isolated by various column chromatographic methods. The structures were identified by spectral data. Eight triterpenoid compounds were isolated and identified as ursolic acid (1), lupeol (2), alpha-amyrin (3), uvaol (4), 3beta-hydroxyurs-11-ene-13beta-olide (5), pomolic acid (6), lup-20 (29)-ene-3beta, 24-diol (7), 3beta, 23-dihydroxy-urs-12-en-28-oic acid (8). The eight compounds were obtained from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U6628
Uvaol, ≥95%
C30H50O2