EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Ring inversion, structural stability and vibrational assignments of sulfolane c-C4H8SO2 and 3-sulfolene c-C4H6SO2.
PMID 18053761
Abstract
The structural stability of sulfolane (tetrahydrothiophene1,1-dioxide) and 3-sulfolene (dihydrothiophene1,1-dioxide) was investigated by DFT-B3LYP and ab initio MP2 calculations with 6-311+G**) basis set. The calculated symmetric ring-puckering potential of 3-sulfolene at the B3LYP level is consistent with a flat minimum that corresponds to a planar ring but at the MP2 level with a double minimum with a low barrier of about 193 cal mol(-1) to ring planarity in reasonable agreement with experimental results. From the calculations at the two levels of theory sulfolane was predicted to exist predominantly in the twist conformation. The vibrational wavenumbers were calculated at the MP2/6-31G** level of theory and the potential energy distributions PED among the symmetry coordinates of the normal modes were computed for the low-energy structure of the molecules. Complete vibrational assignments were provided on the basis of the calculated PED values. The experimental infrared and Raman spectra of the two molecules were compared to the calculated ones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88965
Sulfolane, analytical standard
C4H8O2S
T22209
Sulfolane, 99%
C4H8O2S