EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Preparation of novel beta-cyclodextrin chiral stationary phase based on click chemistry.
PMID 18054027
Abstract
A facile strategy based on click chemistry for preparation of the structurally well-defined native beta-cyclodextrin (beta-CD) based chiral stationary phase (CSP) was proposed. The beta-CD CSP was evaluated by enatioseparation of benzoin, trans-stilbene oxide, Troger's base, bendroflumethiazide, ketoprofen, chlorthalidone, three flavanone compounds and two beta-adrenergic blocking agents under reversed phase high performance liquid chromatography. The chromatographic results demonstrate the chiral separation ability of click beta-CD CSP and illustrate the usefulness of click chemistry in the preparation of beta-CD based CSP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
308323
cis-Stilbene oxide, 97%
C14H12O
S4921
trans-Stilbene oxide, 98%
C14H12O