EMAIL THIS PAGE TO A FRIEND
Journal of Molecular Biology
Mutations in the hydrophobic core of ubiquitin differentially affect its recognition by receptor proteins.
PMID 18054791
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-375
Anti-Ubiquitin Antibody, clone, Upstate®, from rabbit