EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
Effect of combination therapy of a rapid-acting insulin secretagogue (glinide) with premixed insulin in type 2 diabetes mellitus.
PMID 18057760
Abstract
The effect of rapid-acting insulin secretagogues (glinides) on glycemic control when included with insulin therapy for type 2 diabetes remains uncertain. To examine this, we added glinide once a day to twice daily injections of premixed insulin. Seventy-four type 2 diabetic patients, taking twice daily injections of premixed insulin and whose diabetic control was stable, were registered at 6 independent institutions. After a 3-month observation period, 60 patients were administered 10 mg mitiglinide or 90 mg nateglinide at lunchtime without changing their insulin regimen. After 12 weeks, administration of glinide was discontinued and observation was continued. HbA1c levels were measured at the start of glinide administration, after 12 weeks of glinide , and at 12 weeks after discontinuation. HbA1c improved from 7.72+/-0.66% to 7.55+/-0.71% (p <0.01) at Week 12 of glinide administration. Twelve weeks after discontinuation, HbA1c returned to the baseline level (7.72+/-0.81%). This study indicates that the addition of glinide once a day at lunchtime to twice daily injections of premixed insulin is effective for the treatment of type 2 diabetes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0234
Mitiglinide calcium, ≥98% (HPLC)
C19H24NO3·0.5Ca