EMAIL THIS PAGE TO A FRIEND
The Journal of general and applied microbiology
Isolation and characterization of bacteria capable of producing pyridoxamine (PM) and pyridoxamine 5'-phosphate (PMP), vitamin B6 compounds.
PMID 18057820
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82890
Pyridoxamine-5′-phosphate, ≥98.0% (HPLC)
C8H13N2O5P