EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Tailor-made fructooligosaccharides by a combination of substrate and genetic engineering.
PMID 18058889
Abstract
The combination of sucrose analogues as novel substrates (substrate engineering) and highly active recombinant beta-fructofuranosidase from A. niger (genetic engineering) provides a new powerful tool for the efficient preparative synthesis of tailor-made saccharides of the important 1-kestose and 1-nystose type headed with different monosaccharides of interest. These novel compounds have been isolated. They did not display toxic effects or suppress cell growth in initial studies, making these new compounds potential candidates for prebiotics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72555
1-Kestose, analytical standard
C18H32O16