EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Laboratory evolved biocatalysts for stereoselective syntheses of substituted benzaldehyde cyanohydrins.
PMID 18058961
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116025
Mandelonitrile, technical grade
C8H7NO