EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
Potential ipriflavone and warfarin interaction.
PMID 18060912
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
381551
7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one, 97%
C18H16O3