EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Unexpected formation of complex bridged tetrazoles via intramolecular 1,3-dipolar cycloaddition of 1,2-O-cyanoalkylidene derivatives of 3-azido-3-deoxy-D-allose.
PMID 18062945
Abstract
An unexpected and interesting intramolecular side reaction occurred during the attempted synthesis of glycosyl cyanides upon treatment of 1-O-acetyl-3-azido-3-deoxyallose derivatives with TMSCN and different Lewis acids. Exo-1,2-O-cyanoalkylidene derivatives formed by neighboring group participation and attack of cyanide underwent, after Lewis-acid mediated isomerization to the endo-isomer, intramolecular azide-cyanide cycloaddition leading to the formation of tetrazoles embedded in bridged tetracyclic ring systems. The efficiency of cycloaddition is dependent on the ring structure of the sugar (pyranose or furanose). Of the studied molecules, 3-azido-1,2-O-cyanoethylidene-3-deoxy-allopyranose provides the most suitable scaffold for intramolecular [2+3] cycloaddition under exceptionally mild conditions. Our results highlight the capability of carbohydrates to act as scaffolds for the precise positioning of functional groups productive for a specific chemical reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73328
Trimethylsilyl cyanide, technical, ≥95% (GC)
C4H9NSi
212849
Trimethylsilyl cyanide, 98%
C4H9NSi