EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Sequence-specific recognition of double-stranded DNA by synthetic minor groove binder conjugates. toward the construction of artificial site-specific deoxyribonucleases.
PMID 18066826
Abstract
Bis-conjugates of hairpin N-methylpyrrole/N-methylimidazole oligocarboxamide minor groove binders (MGB) possessing enhanced affinity and sequence-specificity for dsDNA were synthesized. Two hairpin MGBs were connected by their N-termini via an aminodiacetate linker. The binding of bis-MGB conjugates to the target DNA was studied by gel mobility retardation, footprinting, and circular dichroism; their affinity and binding mode in the DNA minor groove were determined. In order to functionalize the bis-MGB conjugates, DNA-cleaving agents such as phenanthroline or bipyridine were attached. Effective site-specific cleavage of target DNA in the presence of Cu(2+) ions was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M78801
N-Methylpyrrole, 99%
C5H7N