EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Asymmetric synthesis of novel apio carbocyclic nucleoside analogues as potential antiviral and antitumor agent.
PMID 18066888
Abstract
Novel apio carbocyclic nucleosides 18-21 were asymmetrically synthesized as potential antiviral and antitumor agent, starting from D-ribose employing aldol reaction, RCM reaction and Mitsunobu reaction as key reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109789
2-Amino-6-chloropurine, ≥99%
C5H4ClN5
342300
2-Amino-6-chloropurine, 97%
C5H4ClN5