EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Effect and mechanism of beta-L-D4A on DNA polymerase alpha.
PMID 18069767
Abstract
To investigate the safety of beta-L-D4A on DNA polymerase alpha. Ion exchange chromatography was used to separate DNA polymerase alpha from crude extract of human Hela cells. Detailed kinetic parameters were determined for beta-L-D4A against DNA polymerase alpha. DNA polymerase alpha was purified with 4% yield and 31000 units/mg specific activity. The Michaelis constant (Km = 3.22 micromol/L), 50% inhibition concentration (IC50 = 178.49 micromol/L) and inhibition constant (Ki = 126 micromol/L) of beta-L-D4A were determined by kinetic analysis. beta-L-D4A is a more safe nucleoside for hepatitis B virus infection with a lower host toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1285
2′,3′-Dideoxyadenosine, ≥97% (HPLC)
C10H13N5O2