EMAIL THIS PAGE TO A FRIEND
The journal of headache and pain
Headache caused by handling the pesticide Antracol Combi WP 76: a case report.
PMID 18074103
Abstract
We report on headache caused by the pesticide Antracol Combi WP 76. A headache caused by handling of pesticides should be distinguishable from migraine and headaches from other causes. This 29-year-old woman, who handled pesticides, presented with very strong headache with nausea and vomiting. Poisoning by this pesticide cannot be proven toxicologically, and there is no antidote. Following isolation from the pesticide and abundant hydration, the pain gradually decreased. History, clinical signs and neuroradiological examinations excluded other possible causes of the headache. In treatment of patients in rural areas, attention should also be paid to the pesticide Antracol Combi WP 76 as a possible cause of headaches.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45643
Propineb, PESTANAL®, analytical standard