EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Comment on "1,1,2,2-Tetrachloroethane reactions with OH-, Cr(II), granular iron, and a copper-iron bimetal: insights from product formation and associated carbon isotope fractionation".
PMID 18075113