EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Structural modification of an angiogenesis inhibitor discovered from traditional Chinese medicine and a structure-activity relationship study.
PMID 18078313
Abstract
Pseudolaric acid B (PAB), discovered as a promising angiogensis inhibitor, was served as the anticancer drug lead, and a series of its derivatives were synthesized. Among them, some derivatives, such as 13c- 13k, exhibited potent inhibition on the HMEC-1 cell proliferation and strong cytotoxic activities against the tested six tumor cell lines. The PAB derivatives 13c- 13k also showed significant and specific inhibition on HMEC-1 cell migration in vitro, and only 13d expressed moderate activity against HMEC-1 cell tube formation. The in vitro anticancer tests of the selected natural PAB analogs and the structurally modified PAB derivatives have led to the establishment of a clear structure-activity relationship.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8170 Pseudolaric Acid B, ≥98% (HPLC)
C23H28O8