EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Assimilation of homotaurine-nitrogen by Burkholderia sp. and excretion of sulfopropanoate.
PMID 18081842
Abstract
Homotaurine (3-aminopropanesulfonate), free or derivatized, is in widespread pharmaceutical and laboratory use. Studies with enrichment cultures indicated that the compound is degradable as a sole source of carbon or as a sole source of nitrogen for bacterial growth. A pure culture of Burkholderia sp. was isolated which assimilated the amino group from homotaurine in a glucose-salts medium, and which released an organosulfonate, 3-sulfopropanoate, into the medium stoichiometrically. The deamination involved an inducible 2-oxoglutarate-dependent aminotransferase to yield glutamate, and 3-sulfopropanal. Release of the amino group was attributed to the measured NADP-coupled glutamate dehydrogenase.