EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Diode array detection and simultaneous quantitation of the coeluting atenolol-related synthetic route impurities, PPA-Diol.
PMID 1808216
Abstract
The diode array multichannel detector is used to acquire spectral information at specified intervals in the elution profile of atenolol and its related impurities for post-chromatographic data analysis. The applicability of a number of peak homogeneity testing methods, including spectral normalization, absorbance ratio, chromatographic derivatives, and spectral suppression (SS), are assessed for suitability in simultaneous determinations of the coeluting atenolol-related synthetic impurities, PPA-Diol. Spectral suppression displays a superior performance in comparison to all the other techniques in that both qualitative and quantitative information are acquired on the system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
387738
4-Hydroxyphenylacetamide, 99%
C8H9NO2