EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Conformational analyses of thiirane-based gelatinase inhibitors.
PMID 18083555
Abstract
SB-3CT is a thiirane-containing inhibitor of the gelatinase class of matrix metalloprotease enzymes. In support of the mechanistic study of this inhibition, the conformational analyses of SB-3CT (and of two methyl-substituted derivatives) were undertaken using X-ray crystallography and molecular dynamics simulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03910
Ethylene sulfide, purum, ≥97.5% (GC)
C2H4S
128252
Ethylene sulfide, 98%
C2H4S