EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Synthesis of alpha-ketoamides by a molecular-sieves-promoted formal oxidative coupling of aliphatic aldehydes with isocyanides.
PMID 18092314
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194751
N-tert-Butylhydroxylamine hydrochloride, ≥98%
C4H12ClNO