Molecular biology of the cell
A novel regulatory mechanism of myosin light chain phosphorylation via binding of 14-3-3 to myosin phosphatase.
PMID 18094049
Myosin II phosphorylation-dependent cell motile events are regulated by myosin light-chain (MLC) kinase and MLC phosphatase (MLCP). Recent studies have revealed myosin phosphatase targeting subunit (MYPT1), a myosin-binding subunit of MLCP, plays a critical role in MLCP regulation. Here we report the new regulatory mechanism of MLCP via the interaction between 14-3-3 and MYPT1. The binding of 14-3-3beta to MYPT1 diminished the direct binding between MYPT1 and myosin II, and 14-3-3beta overexpression abolished MYPT1 localization at stress fiber. Furthermore, 14-3-3beta inhibited MLCP holoenzyme activity via the interaction with MYPT1. Consistently, 14-3-3beta overexpression increased myosin II phosphorylation in cells. We found that MYPT1 phosphorylation at Ser472 was critical for the binding to 14-3-3. Epidermal growth factor (EGF) stimulation increased both Ser472 phosphorylation and the binding of MYPT1-14-3-3. Rho-kinase inhibitor inhibited the EGF-induced Ser472 phosphorylation and the binding of MYPT1-14-3-3. Rho-kinase specific siRNA also decreased EGF-induced Ser472 phosphorylation correlated with the decrease in MLC phosphorylation. The present study revealed a new RhoA/Rho-kinase-dependent regulatory mechanism of myosin II phosphorylation by 14-3-3 that dissociates MLCP from myosin II and attenuates MLCP activity.