EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Pyrazines and related compounds; a new synthesis of hydroxypyrazines.
PMID 18108946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
526444
2,4,5-Trimethylbenzoic acid, 97%
C10H12O2