EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
The partial methylation of methyl beta-D-xylopyranoside with methyl sulfate.
PMID 18113533
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5878
Methyl β-D-xylopyranoside, ≥99% (GC)
C6H12O5