EMAIL THIS PAGE TO A FRIEND
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.)
Relation of oxygen and temperature in the preservation of tissues by refrigeration.
PMID 18134009
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63138
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
MgO4S · 7H2O