EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Trends in speciation analysis of vanadium in environmental samples and biological fluids--a review.
PMID 18155404
Abstract
A comprehensive review is presented addressing recent trends in the speciation and determination of vanadium in environmental and biological sample matrices, including important analytical aspects such as sample clean up, pre-concentration and method development. Methodology based on both separation and spectroscopic techniques for the determination of vanadium speciation is discussed. A brief outline of analytical principles, together with an overview of the recent developments and applications of vanadium speciation determination is included. The newer methods for detecting metal ions including hyphenated spectroscopic techniques and sample preparation schemes are also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF67217430
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V
GF58606409
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF26068855
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
V
GF78288946
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
V
GF08542975
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
V
GF02179672
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF88432432
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
V
GF67872213
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
V
GF51153724
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V