EMAIL THIS PAGE TO A FRIEND
Prostaglandins & other lipid mediators
PGF2alpha-associated vascular smooth muscle hypertrophy is ROS dependent and involves the activation of mTOR, p70S6k, and PTEN.
PMID 18160324
Abstract
Prostaglandin F2alpha (PGF2alpha) increases reactive oxygen species (ROS) and induces vascular smooth muscle cell (VSMC) hypertrophy by largely unknown mechanism(s). To investigate the signaling events governing PGF2alpha-induced VSMC hypertrophy we examined the ability of the PGF2alpha analog, fluprostenol to elicit phosphorylation of Akt, the mammalian target of rapamycin (mTOR), ribosomal protein S6 kinase (p70S6k), glycogen synthase kinase-3beta (GSK-3beta), phosphatase and tensin homolog (PTEN), extracellular signal-regulated kinase 1/2 (ERK1/2) and Jun N-terminal kinase (JNK) in growth arrested A7r5 VSMC. Fluprostenol-induced hypertrophy was associated with increased ROS, mTOR translocation from the nucleus to the cytoplasm, along with Akt, mTOR, GSK-3beta, PTEN and ERK1/2 but not JNK phosphorylation. Whereas inhibition of phosphatidylinositol 3-kinase (PI3K) by LY-294002 blocked fluprostenol-induced changes in total protein content, pre-treatment with rapamycin or with the MEK1/2 inhibitor U0126 did not. Taken together, these findings suggest that fluprostenol-induced changes in A7r5 hypertrophy involve mTOR translocation and occur through PI3K-dependent mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8549
Fluprostenol, 10 mg/mL in ethanol, 98%
C23H29F3O6