EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Simultaneous determination of chrysoidine and auramine O in bean products by HPLC].
PMID 18161341
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80028
Chrysoidine G, analytical standard
C12H12N4 · HCl
27240
Chrysoidine G, for microscopy (Bact., Bot., Vit.)
C12H12N4 · HCl