EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Total synthesis and structure assignment of the anthrone C-glycoside cassialoin.
PMID 18163372
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
319899
Anthrone, ACS reagent, 97%
C14H10O