EMAIL THIS PAGE TO A FRIEND
The British journal of ophthalmology
CORTICOSTEROIDS AND FIBROLYSIN IN THE PREVENTION OF LACRIMAL DUCT OBSTRUCTION.
PMID 18170750
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108804
N-Allylthiourea, 98%
C4H8N2S