EMAIL THIS PAGE TO A FRIEND
Indian pediatrics
Baboon syndrome.
PMID 18175852
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7020
Cefadroxil, analytical standard
C16H17N3O5S