EMAIL THIS PAGE TO A FRIEND
Journal of clinical orthodontics : JCO
A non-invasive method for ending thumb- and fingersucking habits.
PMID 18175885
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5765
Denatonium benzoate, ≥98%
C28H34N2O3