EMAIL THIS PAGE TO A FRIEND
Journal of experimental botany
Maize nitrilases have a dual role in auxin homeostasis and beta-cyanoalanine hydrolysis.
PMID 18182427
Abstract
The auxin indole-3-acetic acid (IAA), which is essential for plant growth and development, is suggested to be synthesized via several redundant pathways. In maize (Zea mays), the nitrilase ZmNIT2 is expressed in auxin-synthesizing tissues and efficiently hydrolyses indole-3-acetonitrile to IAA. Zmnit2 transposon insertion mutants were compromised in root growth in young seedlings and sensitivity to indole-3-acetonitrile, and accumulated lower quantities of IAA conjugates in kernels and root tips, suggesting a substantial contribution of ZmNIT2 to total IAA biosynthesis in maize. An additional enzymatic function, turnover of beta-cyanoalanine, is acquired when ZmNIT2 forms heteromers with the homologue ZmNIT1. In plants carrying an insertion mutation in either nitrilase gene this activity was strongly reduced. A dual role for ZmNIT2 in auxin biosynthesis and in cyanide detoxification as a heteromer with ZmNIT1 is therefore proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129453
3-Indoleacetonitrile, 98%
C10H8N2
C9650
β-Cyano-L-alanine, ≥95%
C4H6N2O2