EMAIL THIS PAGE TO A FRIEND
European journal of mass spectrometry (Chichester, England)
Copper-induced oligomerization of peptides: a model study.
PMID 18192726
Abstract
In this work, copper-binding of the tetraglycine peptide (Gly-Gly-Gly-Gly) was studied by electrospray ionization mass spectrometry. Experiments were performed under alkaline conditions, in the presence of ethanolamine (pH 10.95). We observed that the presence of copper(II) ions induces the aggregation of the peptide and the formation of copper-bound complexes with higher molecular mass is favored, such as the oligomer complexes [3M+2Cu-3H](+) and [4M+3Cu-5H](+). At 1:1 peptide-copper(II) ion ratio, the singly charged [3M+2Cu-3H](+) oligomer complex is the base peak in the mass spectrum. Metal ion-induced oligomer-ization of neurotoxic peptides is well known in the literature; however, there are very few examples in which such oligomerization was directly observed by mass spectrometry. Our results show that application of short peptides can be useful to study the -mechanism of metal ion binding and metal ion-induced oligomerization of peptides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3882
Gly-Gly-Gly-Gly
C8H14N4O5