EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
New trisubstituted 1,2,4-triazole derivatives as potent ghrelin receptor antagonists. 3. Synthesis and pharmacological in vitro and in vivo evaluations.
PMID 18193826
Abstract
Ghrelin receptor ligands based on trisubstituted 1,2,4-triazole structure were synthesized and evaluated for their in vitro binding and biological activity. In this study, we explored the replacement of the alpha-aminoisobutyryl moiety by aromatic or heteroaromatic groups. Compounds 5 and 34 acted as potent in vivo antagonists of hexarelin-stimulated food intake. These two compounds did not stimulate growth hormone secretion in rodents and did not antagonize growth hormone secretion induced by hexarelin.