Journal of physiology and pharmacology : an official journal of the Polish Physiological Society
DDT- and DDE-induced disruption of ovarian steroidogenesis in prepubertal porcine ovarian follicles: a possible interaction with the main steroidogenic enzymes and estrogen receptor beta.
PMID 18195494
We evaluated impact of DDT isomers, o, p'- DDT [1, 1-dichloro-2, 2-bis (p, p'-chlorophenyl) ethylene] and p, p'-DDT [1, 1, 1-trichloro-2, 2-bis (p-chlorophenyl) ethane], and their metabolites, o, p'-DDE and p, p'-DDE, on ovarian steroidogenesis. All these compounds, except for p, p'-DDT, demonstrated estrogenic effects on steroid secretion in co-cultures of porcine prepubertal granulosa and theca cells. p,p'-DDT decreased progesterone and estradiol release, which was reversed by the addition of testosterone. In contrast, o, p'-DDT inhibited progesterone secretion with parallel stimulation of basal and testosterone-stimulated estradiol release. DDEs stimulated progesterone and estradiol secretion. The fluorometric assay confirmed that p,p'-DDE, o,p'-DDT, and o,p'-DDE stimulated aromatase activity. Western blots indicated that o,p-DDT and o,p'-DDE diminished the expression of estrogen receptor beta (ERbeta). This study demonstrated the isomer-dependent action of DDT in pig ovarian cells. We propose that DDT could disrupt ovarian steroidogenesis either by interfering with main steroidogenic enzymes or affecting ERbeta.