EMAIL THIS PAGE TO A FRIEND
Tetrahedron
Palladium-catalyzed cross-coupling reaction of alkenyl bromides with potassium alkyltrifluoroborates.
PMID 18196177
Abstract
The Suzuki-Miyaura-type cross-coupling reaction of potassium alkyltrifluoroborates with various alkenyl bromides in the presence of 10 mol % of PdCl(2)(dppf)·CH(2)Cl(2) and 3.0 equiv of Cs(2)CO(3) in aqueous toluene at 80 °C provided the desired compounds in 49-95% yields. A variety of functional groups in the potassium alkyltrifluoroborates were tolerated under the basic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
371289
2-Bromo-3-methyl-2-butene, 96%
C5H9Br
453595
2-Bromo-3-methyl-2-cyclopenten-1-one, 98%
C6H7BrO