EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Total synthesis of 2-hydroxytetracosanolide and 2-hydroxy-24-oxooctacosanolide by using an effective lactonization.
PMID 18203212
Abstract
We have developed effective methods for the total synthesis of 2-hydroxytetracosanolide and 2-hydroxy-24-oxooctacosanolide, the defensive salivary secretions of termites. The former natural product isolated from Armitermes neotenicus, a species of termite that inhabits Guyana, contains a 25-membered lactone backbone, and the latter, extracted from Pseudacanthoterme springer, an African termite, includes a 29-membered lactone moiety. The key cyclization to produce the 25- or 29-membered lactone core is performed by using 2-methyl-6-nitrobenzoic anhydride (MNBA) with a stoichiometric amount of 4-(dimethylamino)pyridine (DMAP) or a catalytic amount of 4-(dimethylamino)pyridine N-oxide (DMAPO).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
385980
Benzoic anhydride, ≥95%
C14H10O3