EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Synthesis and quantitative evaluation of Glycero-D-manno-heptose binding to concanavalin A by fluorous-tag assistance.
PMID 18203223