EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Regio- and enantioselective o-allylation of phenol and alcohol catalyzed by a planar-chiral cyclopentadienyl ruthenium complex.
PMID 18205149