EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Proline bis-amides as potent dual orexin receptor antagonists.
PMID 18207395
Abstract
A series of OX(2)R/OX(1)R dual orexin antagonists was prepared based on a proline bis-amide identified as a screening lead. Through a combination of classical and library synthesis, potency enhancing replacements for both amide portions were discovered. N-methylation of the benzimidazole moiety within the lead structure significantly reduced P-gp susceptibility while increasing potency, giving rise to good brain penetration. A compound from this series has demonstrated in vivo central activity when dosed peripherally in a pharmacodynamic model of orexin activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2117
TCS-1102, ≥98% (HPLC)
C27H26N4O2S