EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Facile assembly of hybrid materials containing polyoxometalate cluster anions and organic dye cations: crystal structures and initial spectral characterization.
PMID 18209794
Abstract
Salts of the pararosaniline dye cation and four polyoxometalate cluster anions have been isolated under both ambient and hydrothermal conditions; structural and initial spectroscopic data are consistent with significant perturbation of ion electronic states induced by charge-assisted N-H---O hydrogen bonds.