EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Oxidation of primary alcohols to methyl esters by hydrogen transfer.
PMID 18209810
Abstract
The oxidation of alcohols in the presence of methanol has been achieved using a ruthenium catalyst with crotononitrile as the hydrogen acceptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252522
Crotononitrile, mixture of cis and trans, 99%
C4H5N