EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Genomewide screening for genes associated with gliotoxin resistance and sensitivity in Saccharomyces cerevisiae.
PMID 18212113
Abstract
Gliotoxin (GT) is a secondary fungal metabolite with pleiotropic immunosuppressive properties that have been implicated in Aspergillus virulence. However, the mechanisms of GT cytotoxicity and its molecular targets in eukaryotic cells have not been fully characterized. We screened a haploid library of Saccharomyces cerevisiae single-gene deletion mutants (4,787 strains in EUROSCARF) to identify nonessential genes associated with GT increased resistance (GT-IR) and increased sensitivity (GT-IS). The susceptibility of the wild-type parental strain BY4741 to GT was initially assessed by broth microdilution methods using different media. GT-IR and GT-IS were defined as a fourfold increase and decrease, respectively, in MIC, and this was additionally confirmed by susceptibility testing on agar yeast extract-peptone-glucose plates. The specificity of GT-IR and GT-IS mutants exhibiting normal growth compared with the wild-type strain was further tested in studies of their susceptibility to conventional antifungal agents, cycloheximide, and H2O2. GT-IR was associated with the disruption of genes acting in general metabolism (OPI1, SNF1, IFA38), mitochondrial function (RTG2), DNA damage repair (RAD18), and vesicular transport (APL2) and genes of unknown function (YGL235W, YOR345C, YLR456W, YGL072C). The disruption of three genes encoding transsulfuration (CYS3), mitochondrial function (MEF2), and an unknown function (YKL037W) led to GT-IS. Specificity for GT-IR and GT-IS was observed in all mutants. Importantly, the majority (69%) of genes implicated in GT-IR (6/10) and GT-IS (2/3) have human homologs. We identified novel Saccharomyces genes specifically implicated in GT-IR or GT-IS. Because most of these genes are evolutionarily conserved, further characterization of their function could improve our understanding of GT cytotoxicity mechanisms in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9893
Gliotoxin from Gliocladium fimbriatum
C13H14N2O4S2