EMAIL THIS PAGE TO A FRIEND
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993)
[Interferonogenic activity of amixin analogs and diphenyl derivatives].
PMID 18217355
Abstract
Properties of interferon productivity of several inducers of interferon, analogs of amixin, have been studied in vivo. It has been shown, that under the influence of injected agents the content of endogenic interferon in the blood serum of laboratory animals increased and their non-fractionated cells of the peripheral blood (splenocites and macrophages) synthesized alpha- and gamma-interferon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220957
Tilorone dihydrochloride, 95%
C25H34N2O3 · 2HCl