EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Magnesium absorption by lactating dairy cows on a grass silage-based diet supplied with different potassium and magnesium levels.
PMID 18218762
Abstract
The objective of the present study was to investigate the interactions of dietary K intake typical for forage-based diets on Mg balance in lactating dairy cows. Six lactating multiparous cows of the Swedish Red and White breed in midlactation were used. Two concentrations of Mg (1.9 and 4.3 g/kg of dry matter) and 3 concentrations of K (19, 28, and 37 g of K/kg of dry matter) were obtained by adding appropriate amounts of MgO and KHCO(3) to the diet. The experimental setup was a 6 x 6 Latin square design with a 2 x 3 factorial arrangement of treatments. Each experimental period lasted 14 d (9-d treatment adaptation period and 5-d data collection). There was no effect of Mg or K dietary supplementation on milk yield. Supplementing the ration with K did not significantly affect the Mg apparent absorption, urinary Mg excretion, or plasma Mg concentration. The Mg balance, estimated as the Mg losses in milk and urine, was positively related to Mg intake but not affected by K intake. The amount of apparently digested Mg was related to the Mg balance. The apparent digestibility ranged from 0.12 to 0.24 with no effect of mineral supplementation. There was a significant curvilinear relationship between plasma Mg and urinary Mg excretion, with a more marked increase in urinary Mg excretion at higher plasma levels of Mg.