EMAIL THIS PAGE TO A FRIEND
Nature materials
Complex thermoelectric materials.
PMID 18219332
Abstract
Thermoelectric materials, which can generate electricity from waste heat or be used as solid-state Peltier coolers, could play an important role in a global sustainable energy solution. Such a development is contingent on identifying materials with higher thermoelectric efficiency than available at present, which is a challenge owing to the conflicting combination of material traits that are required. Nevertheless, because of modern synthesis and characterization techniques, particularly for nanoscale materials, a new era of complex thermoelectric materials is approaching. We review recent advances in the field, highlighting the strategies used to improve the thermopower and reduce the thermal conductivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
776696
Germanium telluride, 99.999% trace metals basis
GeTe