EMAIL THIS PAGE TO A FRIEND
Current medicinal chemistry
Microtubule-stabilizing natural products as promising cancer therapeutics.
PMID 18220732
Abstract
Paclitaxel and related taxanes exhibit their anticancer activity by promoting tubulin polymerization and stabilizing microtubules, which results in mitotic G2/M arrest and apoptosis. The clinical success of paclitaxel in treating a wide array of tumor types has led to numerous efforts to identify novel natural products with paclitaxel-like mechanisms of action, but which may overcome some of the liabilities of the taxanes. Although the list of natural products that share the paclitaxel-like mechanism is relatively small, it continues to expand and currently includes a number of structurally distinct classes. Despite the mechanistic similarities between these classes, differences exist which may translate into their differential efficacy in the clinic. The past several years have seen a considerable amount of pre-clinical and clinical progress in developing these novel microtubule-stabilizing natural products as cancer therapeutics. This review focuses primarily on recent advances published since 2002.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7402
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
C47H51NO14